Assay Details
Gene Symbol: Adcy9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767525.1 | XP_008765747.1 | 2 - 3 | 1903 | 104 | |
XM_008767526.1 | XP_008765748.1 | 2 - 3 | 1903 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
