Assay Details
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009619.1 | NP_001009619.1 | 10 - 11 | 930 | 106 | |
GenBank mRNA | BC088221.1 | - | 10 - 11 | 930 | 106 | |
FQ214074.1 | - | 9 - 10 | 874 | 106 | ||
FQ218444.1 | - | 10 - 11 | 943 | 106 | ||
FQ220638.1 | - | 10 - 11 | 943 | 106 | ||
FQ222868.1 | - | 10 - 11 | 947 | 106 | ||
FQ229330.1 | - | 10 - 11 | 946 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
