Assay Details
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009619.1 | NP_001009619.1 | 4 - 5 | 350 | 92 | |
GenBank mRNA | BC088221.1 | - | 4 - 5 | 350 | 92 | |
FQ218444.1 | - | 4 - 5 | 363 | 92 | ||
FQ220638.1 | - | 4 - 5 | 92 | |||
FQ222868.1 | - | 4 - 5 | 367 | 92 | ||
FQ229330.1 | - | 4 - 5 | 366 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
