Assay Details
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009619.1 | NP_001009619.1 | 6 - 7 | 478 | 77 | |
GenBank mRNA | BC088221.1 | - | 6 - 7 | 478 | 77 | |
FQ214074.1 | - | 5 - 6 | 422 | 77 | ||
FQ218444.1 | - | 6 - 7 | 491 | 77 | ||
FQ220638.1 | - | 6 - 7 | 491 | 77 | ||
FQ222868.1 | - | 6 - 7 | 495 | 77 | ||
FQ229330.1 | - | 6 - 7 | 494 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
