Assay Details
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009619.1 | NP_001009619.1 | 7 - 8 | 629 | 137 | |
GenBank mRNA | BC088221.1 | - | 7 - 8 | 629 | 137 | |
FQ214074.1 | - | 6 - 7 | 573 | 137 | ||
FQ218444.1 | - | 7 - 8 | 642 | 137 | ||
FQ222868.1 | - | 7 - 8 | 646 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
