Assay Details
Gene Symbol: Atf7ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017471.1 | NP_001017471.1 | 6 - 7 | 725 | 169 | |
XM_006245728.2 | XP_006245790.1 | 6 - 7 | 879 | 169 | ||
XM_006245732.2 | XP_006245794.1 | 6 - 7 | 845 | 169 | ||
XM_006245735.1 | XP_006245797.1 | 4 - 5 | 453 | 169 | ||
XM_008767495.1 | XP_008765717.1 | 6 - 7 | 880 | 169 | ||
XM_008767496.1 | XP_008765718.1 | 6 - 7 | 878 | 169 | ||
GenBank mRNA | BC083663.1 | - | 6 - 7 | 725 | 169 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
