Assay Details
Gene Symbol: Naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014226.2 | NP_001014248.2 | 2 - 3 | 210 | 112 | |
XM_008767519.1 | XP_008765741.1 | 1 - 2 | 108 | 112 | ||
XM_008767520.1 | XP_008765742.1 | 2 - 3 | 246 | 112 | ||
GenBank mRNA | AY316589.1 | - | 2 - 3 | 252 | 112 | |
BC105304.1 | - | 2 - 3 | 210 | 112 | ||
FQ212054.1 | - | 2 - 3 | 214 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
