Assay Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001071128.5 | XP_001071128.2 | 3 - 4 | 1042 | 151 | |
XM_006236596.2 | XP_006236658.1 | 3 - 4 | 1042 | 151 | ||
XM_006236597.2 | XP_006236659.1 | 3 - 4 | 1043 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
