Assay Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001071128.5 | XP_001071128.2 | 8 - 9 | 3548 | 76 | |
XM_006236596.2 | XP_006236658.1 | 8 - 9 | 3545 | 76 | ||
XM_006236597.2 | XP_006236659.1 | 9 - 10 | 3546 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
