Assay Details
Gene Symbol: Snca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236591.2 | XP_006236653.1 | 4 - 5 | 749 | 75 | |
GenBank mRNA | S73008.1 | - | 4 - 5 | 399 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
