Assay Details
Gene Symbol: Mad2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106594.1 | NP_001100064.1 | 1 - 2 | 191 | 75 | |
XM_006236619.2 | XP_006236681.1 | 1 - 2 | 281 | 75 | ||
XM_006236620.2 | XP_006236682.1 | 1 - 2 | 281 | 75 | ||
XM_006236621.2 | XP_006236683.1 | 1 - 2 | 281 | 75 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
