Assay Details
Gene Symbol: Mad2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106594.1 | NP_001100064.1 | 4 - 5 | 561 | 108 | |
XM_006236619.2 | XP_006236681.1 | 4 - 5 | 651 | 108 | ||
XM_006236620.2 | XP_006236682.1 | 4 - 5 | 651 | 108 | ||
XM_006236621.2 | XP_006236683.1 | 4 - 5 | 651 | 108 | ||
GenBank mRNA | FQ222936.1 | - | 4 - 5 | 537 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
