Assay Details
Gene Symbol: Mad2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236621.2 | XP_006236683.1 | 5 - 6 | 829 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
