Assay Details
Gene Symbol: Il23r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001072576.5 | XP_001072576.3 | 8 - 9 | 1152 | 100 |
Gene Symbol: LOC103690079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236622.2 | XP_006236684.2 | 1 - 2 | 1442 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006236622.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
