Assay Details
Gene Symbol: LOC103692165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763034.1 | XP_008761256.1 | 3 - 4 | 2947 | 125 | |
XM_008763035.1 | XP_008761257.1 | 3 - 4 | 874 | 125 | ||
XM_008763036.1 | XP_008761258.1 | 3 - 4 | 711 | 125 | ||
XM_008763037.1 | XP_008761259.1 | 3 - 4 | 2947 | 125 | ||
XM_008763038.1 | XP_008761260.1 | 3 - 4 | 2947 | 125 |
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_184046.1 | NP_908935.1 | 919 | 125 | ||
XM_006236729.2 | XP_006236791.1 | 2947 | 125 | |||
XM_006236731.2 | XP_006236793.1 | 2947 | 125 | |||
XM_006236732.2 | XP_006236794.1 | 2947 | 125 | |||
XM_008763054.1 | XP_008761276.1 | 874 | 125 | |||
XM_008763055.1 | XP_008761277.1 | 711 | 125 | |||
GenBank mRNA | AY348867.1 | - | 919 | 125 | ||
AY348868.1 | - | 919 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
