Assay Details
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037768.1 | NP_001032857.1 | 1 - 2 | 62 | 91 | |
XM_006236740.2 | XP_006236802.1 | 1 - 2 | 1118 | 91 | ||
GenBank mRNA | BC079456.1 | - | 1 - 2 | 44 | 91 | |
BC107647.1 | - | 1 - 2 | 62 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC107647.1, NM_001037768.1, XM_006236740.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
