Assay Details
Gene Symbol: Cnbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022598.1 | NP_072120.1 | 1 - 2 | 154 | 134 | |
XM_006236854.2 | XP_006236916.1 | 1 - 2 | 170 | 134 | ||
GenBank mRNA | BC062225.1 | - | 1 - 2 | 138 | 6889682 | 134 |
D45254.1 | - | 1 - 2 | 154 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC062225.1,D45254.1,NM_022598.1,XM_006236854.2 transcripts.
This assay is in the 5'UTR region of the BC062225.1,D45254.1,NM_022598.1,XM_006236854.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
