Assay Details
Gene Symbol: Cnbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022598.1 | NP_072120.1 | 4 - 5 | 588 | 115 | |
XM_006236854.2 | XP_006236916.1 | 4 - 5 | 583 | 115 | ||
GenBank mRNA | AB158421.1 | - | 3 - 4 | 423 | 115 | |
AB158422.1 | - | 3 - 4 | 423 | 115 | ||
AF242550.1 | - | 4 - 5 | 474 | 115 | ||
BC062225.1 | - | 4 - 5 | 572 | 6889682 | 115 | |
D45254.1 | - | 4 - 5 | 588 | 115 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
