Assay Details
Gene Symbol: Tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763059.1 | XP_008761281.1 | 6 | 78 | ||
XM_008763060.1 | XP_008761282.1 | 6 | 78 | |||
XM_008763061.1 | XP_008761283.1 | 6 | 78 | |||
GenBank mRNA | FQ218231.1 | - | 6 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
