Assay Details
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729395.4 | XP_002729441.3 | 1 - 2 | 1064 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
