Assay Details
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147177.1 | NP_671706.1 | 779 | 121 | ||
XM_006236865.2 | XP_006236927.1 | 853 | 121 | |||
GenBank mRNA | AB001581.1 | - | 766 | 121 | ||
AB002406.1 | - | 779 | 121 | |||
BC072511.1 | - | 764 | 121 | |||
BC086531.1 | - | 797 | 121 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
