Assay Details
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147177.1 | NP_671706.1 | 1050 | 81 | ||
XM_006236865.2 | XP_006236927.1 | 1124 | 81 | |||
GenBank mRNA | AB001581.1 | - | 1037 | 81 | ||
AB002406.1 | - | 1050 | 81 | |||
BC072511.1 | - | 1035 | 81 | |||
BC086531.1 | - | 1068 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
