Assay Details
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053879.1 | NP_446331.1 | 13 - 14 | 1769 | 112 | |
XM_008763166.1 | XP_008761388.1 | 9 - 10 | 1500 | 112 | ||
GenBank mRNA | U35371.1 | - | 13 - 14 | 1769 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
