Assay Details
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053879.1 | NP_446331.1 | 2 - 3 | 264 | 141 | |
GenBank mRNA | U35371.1 | - | 2 - 3 | 264 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_053879.1, U35371.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
