Assay Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053752.2 | NP_446204.2 | 2 - 3 | 216 | 74 | |
GenBank mRNA | BC061537.1 | - | 2 - 3 | 220 | 74 | |
FQ212668.1 | - | 2 - 3 | 213 | 74 | ||
FQ214532.1 | - | 2 - 3 | 175 | 74 | ||
FQ214959.1 | - | 2 - 3 | 205 | 74 | ||
FQ215028.1 | - | 2 - 3 | 174 | 74 | ||
FQ215469.1 | - | 2 - 3 | 213 | 74 | ||
FQ216893.1 | - | 2 - 3 | 205 | 74 | ||
FQ217244.1 | - | 2 - 3 | 230 | 74 | ||
U75394.1 | - | 2 - 3 | 157 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
