Assay Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053752.2 | NP_446204.2 | 4 - 5 | 549 | 109 | |
XM_008762973.1 | XP_008761195.1 | 3 - 4 | 462 | 109 | ||
GenBank mRNA | BC061537.1 | - | 4 - 5 | 553 | 6920009 | 109 |
FQ212668.1 | - | 4 - 5 | 109 | |||
FQ214532.1 | - | 4 - 5 | 508 | 109 | ||
FQ214959.1 | - | 4 - 5 | 538 | 109 | ||
FQ215028.1 | - | 4 - 5 | 507 | 109 | ||
FQ215469.1 | - | 4 - 5 | 546 | 109 | ||
FQ216893.1 | - | 4 - 5 | 538 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
