Assay Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053752.2 | NP_446204.2 | 5 - 6 | 597 | 105 | |
XM_008762973.1 | XP_008761195.1 | 4 - 5 | 510 | 105 | ||
GenBank mRNA | BC061537.1 | - | 5 - 6 | 601 | 6920009 | 105 |
FQ212668.1 | - | 5 - 6 | 594 | 105 | ||
FQ214532.1 | - | 5 - 6 | 556 | 105 | ||
FQ214959.1 | - | 5 - 6 | 586 | 105 | ||
FQ215028.1 | - | 5 - 6 | 555 | 105 | ||
FQ215469.1 | - | 5 - 6 | 594 | 105 | ||
FQ216893.1 | - | 5 - 6 | 586 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
