Assay Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053752.2 | NP_446204.2 | 6 - 7 | 684 | 126 | |
XM_008762973.1 | XP_008761195.1 | 5 - 6 | 597 | 126 | ||
GenBank mRNA | BC061537.1 | - | 6 - 7 | 688 | 6920009 | 126 |
FQ212668.1 | - | 6 - 7 | 681 | 126 | ||
FQ214959.1 | - | 6 - 7 | 673 | 126 | ||
FQ215028.1 | - | 6 - 7 | 642 | 126 | ||
FQ215469.1 | - | 6 - 7 | 681 | 126 | ||
FQ216893.1 | - | 6 - 7 | 673 | 127 | ||
FQ217244.1 | - | 6 - 7 | 698 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
