Assay Details
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053752.2 | NP_446204.2 | 7 - 8 | 834 | 99 | |
XM_008762973.1 | XP_008761195.1 | 6 - 7 | 747 | 99 | ||
GenBank mRNA | BC061537.1 | - | 7 - 8 | 838 | 99 | |
FQ212668.1 | - | 7 - 8 | 831 | 99 | ||
FQ214532.1 | - | 7 - 8 | 793 | 99 | ||
FQ215028.1 | - | 7 - 8 | 792 | 99 | ||
FQ215469.1 | - | 7 - 8 | 831 | 99 | ||
FQ216893.1 | - | 7 - 8 | 824 | 99 | ||
FQ217244.1 | - | 7 - 8 | 848 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
