Assay Details
Gene Symbol: Rpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108632.1 | NP_001102102.1 | 2 - 3 | 358 | 82 | |
XM_006236628.2 | XP_006236690.1 | 2 - 3 | 356 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
