Assay Details
Gene Symbol: Rpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108632.1 | NP_001102102.1 | 7 - 8 | 746 | 139 | |
XM_006236628.2 | XP_006236690.1 | 6 - 7 | 684 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P139168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12410 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P139168 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P139168 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P139168 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P391726 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 27082 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10863086 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863083 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863081 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863087 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863082 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863088 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863080 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863084 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863085 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10863079 | - | - |
Rat Genome 230 2.0 Array | 1389468_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376559_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376559_at | - | - |
Rat Genome 230 2.0 Array | 1389468_at | - | - |
Affymetrix HT Rat Focus Array | 1389468_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389468_at | - | - |
Affymetrix HT Rat Focus Array | 1376559_at | - | - |
Affymetrix HT Rat Focus Array | 1376559_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400021484.rn.1 | TC0400003039.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021482.rn.1 | TC0400003039.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021485.rn.1 | TC0400003039.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021486.rn.1 | TC0400003039.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021483.rn.1 | TC0400003039.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021481.rn.1 | TC0400003039.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400021480.rn.1 | TC0400003039.rn.1 | RTA TC |
Rat Expression Set 230 | 1389468_PM_at | - | - |
Rat Expression Set 230 | 1376559_PM_at | - | - |
Rat Expression Set 230 | 1376559_at | - | - |
Rat Expression Set 230 | 1389468_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863079 | - | - |
Rat Genome U34 Set | rc_AA996886_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21962819 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375214 | - | - |
