Assay Details
Gene Symbol: Ccdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236885.2 | XP_006236947.1 | 14 - 15 | 2295 | 88 | |
XM_006236886.2 | XP_006236948.1 | 13 - 14 | 2119 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
