Assay Details
Gene Symbol: Ghrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021669.2 | NP_067701.1 | 1 - 2 | 140 | 130 | |
GenBank mRNA | AB029433.1 | - | 1 - 2 | 141 | 130 | |
BF543543.1 | - | 1 - 2 | 136 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
