Assay Details
Gene Symbol: Ghrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021669.2 | NP_067701.1 | 2 - 3 | 249 | 106 | |
GenBank mRNA | AB029433.1 | - | 2 - 3 | 250 | 106 | |
AB035699.1 | - | 2 - 3 | 243 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
