Assay Details
Gene Symbol: Mrpl53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108635.2 | NP_001102105.1 | 2 - 3 | 213 | 71 | |
GenBank mRNA | BC166928.1 | - | 2 - 3 | 213 | 71 |
Gene Symbol: LOC103692169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763084.1 | XP_008761306.1 | 2 - 3 | 483 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
