Assay Details
Gene Symbol: Emc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008355.1 | NP_001008356.1 | 3 - 4 | 413 | 102 | |
GenBank mRNA | BC085846.1 | - | 3 - 4 | 413 | 102 | |
FQ220446.1 | - | 3 - 4 | 298 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
