Assay Details
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109398.1 | NP_001102868.1 | 5 - 6 | 742 | 87 | |
GenBank mRNA | FQ234580.1 | - | 6 - 7 | 794 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
