Assay Details
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109398.1 | NP_001102868.1 | 6 - 7 | 837 | 101 | |
GenBank mRNA | FQ231869.1 | - | 2 - 3 | 148 | 101 | |
FQ234580.1 | - | 7 - 8 | 889 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
