Assay Details
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100750.1 | NP_001094220.1 | 8 - 9 | 922 | 154 | |
GenBank mRNA | BC079367.1 | - | 8 - 9 | 915 | 154 | |
BC089863.1 | - | 7 - 8 | 780 | 154 | ||
BC099746.1 | - | 8 - 9 | 906 | 154 | ||
BC160886.1 | - | 8 - 9 | 922 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
