Assay Details
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106613.2 | NP_001100083.2 | 3 - 4 | 504 | 169 | |
XM_006236962.2 | XP_006237024.1 | 3 - 4 | 578 | 169 | ||
GenBank mRNA | BC167050.1 | - | 3 - 4 | 422 | 169 | |
EV770270.1 | - | 3 - 4 | 523 | 170 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
