Assay Details
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106613.2 | NP_001100083.2 | 7 - 8 | 857 | 83 | |
XM_006236962.2 | XP_006237024.1 | 7 - 8 | 931 | 83 | ||
GenBank mRNA | BC167050.1 | - | 7 - 8 | 775 | 83 | |
EV776660.1 | - | 4 - 5 | 336 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44619 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071396 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071396 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071396 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P568060 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P310658 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64362 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25694 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10857480 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857474 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857477 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857481 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857478 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857475 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857476 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857479 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857482 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10857473 | - | - |
Rat Genome 230 2.0 Array | 1393231_at | - | - |
Rat Genome 230 2.0 Array | 1388865_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388865_at | - | - |
Rat Genome 230 2.0 Array | 1392997_at | - | - |
Rat Genome 230 2.0 Array | 1392997_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393231_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393231_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392997_at | - | - |
Affymetrix HT Rat Focus Array | 1393231_at | - | - |
Affymetrix HT Rat Focus Array | 1388865_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392997_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388865_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400010489.rn.1 | TC0400001497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400010491.rn.1 | TC0400001497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400010490.rn.1 | TC0400001497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400010492.rn.1 | TC0400001497.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400010493.rn.1 | TC0400001497.rn.1 | RTA TC |
Rat Expression Set 230 | 1388865_at | - | - |
Rat Expression Set 230 | 1388865_PM_at | - | - |
Rat Expression Set 230 | 1392997_PM_at | - | - |
Rat Expression Set 230 | 1393231_PM_at | - | - |
Rat Expression Set 230 | 1392997_at | - | - |
Rat Expression Set 230 | 1393231_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10857473 | - | - |
Rat Genome U34 Set | rc_AA848965_at | - | - |
Rat Genome U34 Set | rc_AA964661_at | - | - |
Rat Genome U34 Set | rc_AI235325_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22001793 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363915 | - | - |
