Assay Details
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173382.1 | NP_001166853.1 | 7 - 8 | 1374 | 74 | |
XM_008763076.1 | XP_008761298.1 | 7 - 8 | 1425 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
