Assay Details
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134351.1 | NP_599178.1 | 8 - 9 | 1161 | 147 | |
GenBank mRNA | J05571.1 | - | 8 - 9 | 1161 | 147 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15601 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P027404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162836 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027404 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15601 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027404 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15601 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15601 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P027404 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1037609 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P112050 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15601 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P118993 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16179 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10863223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767002 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863229 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10863221 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10767001 | - | - |
Rat Genome 230 2.0 Array | 1387737_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376289_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371350_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380177_at | - | - |
Rat Genome 230 2.0 Array | 1387737_at | - | - |
Rat Genome 230 2.0 Array | 1371350_at | - | - |
Rat Genome 230 2.0 Array | 1380177_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376289_at | - | - |
Affymetrix HT Rat Focus Array | 1371350_at | - | - |
Affymetrix HT Rat Focus Array | 1387737_at | - | - |
Affymetrix HT Rat Focus Array | 1380177_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380177_at | - | - |
Affymetrix HT Rat Focus Array | 1371350_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387737_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400021830.rn.1 | TC0400003080.rn.1 | RTA TC |
Rat Expression Set 230 | 1371350_at | - | - |
Rat Expression Set 230 | 1371350_PM_at | - | - |
Rat Expression Set 230 | 1387737_at | - | - |
Rat Expression Set 230 | 1387737_PM_at | - | - |
Rat Expression Set 230 | 1380177_at | - | - |
Rat Expression Set 230 | 1380177_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10863221 | - | - |
Rat Genome U34 Set | J05571_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21445584 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359095 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364192 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361968 | - | - |
RatRef-12 Expression BeadChip | ILMN_49623 | - | - |
RatRef-12 Expression BeadChip | ILMN_67451 | - | - |
