Assay Details
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106612.1 | NP_001100082.1 | 6 - 7 | 604 | 155 | |
XM_008763159.1 | XP_008761381.1 | 6 - 7 | 1018 | 155 | ||
XM_008763160.1 | XP_008761382.1 | 8 - 9 | 1088 | 155 | ||
XM_008763161.1 | XP_008761383.1 | 7 - 8 | 952 | 155 | ||
GenBank mRNA | BC168936.1 | - | 6 - 7 | 591 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P466595 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P526712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96711 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P466595 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P526712 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P526712 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P466595 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P466595 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P526712 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466595 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P206290 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69813 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2909 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58241 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2198 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10864428 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864429 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864430 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864431 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864432 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864427 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864426 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10864425 | - | - |
Rat Genome 230 2.0 Array | 1390396_at | - | - |
Rat Genome 230 2.0 Array | 1391935_at | - | - |
Rat Genome 230 2.0 Array | 1391935_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390396_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391935_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390396_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391935_at | - | - |
Affymetrix HT Rat Focus Array | 1390396_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400024358.rn.1 | TC0400003446.rn.1 | RTA TC |
Rat Expression Set 230 | 1390396_at | - | - |
Rat Expression Set 230 | 1390396_PM_at | - | - |
Rat Expression Set 230 | 1391935_at | - | - |
Rat Expression Set 230 | 1391935_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10864425 | - | - |
Rat Genome U34 Set | rc_AI101338_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21974333 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365630 | - | - |
