Assay Details
Gene Symbol: Nfu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106606.2 | NP_001100076.2 | 6 - 7 | 543 | 106 | |
XM_008763063.1 | XP_008761285.1 | 6 - 7 | 776 | 106 | ||
GenBank mRNA | FM064033.1 | - | 5 - 6 | 369 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
