Assay Details
Gene Symbol: Hmces | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025047.1 | NP_001020218.1 | 1 - 2 | 211 | 134 | |
XM_006236851.2 | XP_006236913.1 | 2 - 3 | 595 | 134 | ||
GenBank mRNA | BC083690.1 | - | 1 - 2 | 211 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
