Assay Details
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006978.1 | NP_001006979.1 | 2 - 3 | 87 | 152 | |
GenBank mRNA | BC084705.1 | - | 2 - 3 | 87 | 152 | |
FQ216715.1 | - | 2 - 3 | 105 | 152 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC084705.1, NM_001006978.1 transcripts.
This assay is in the 5'UTR region of the BC084705.1, NM_001006978.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
