Assay Details
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006978.1 | NP_001006979.1 | 3 - 4 | 192 | 66 | |
GenBank mRNA | BC084705.1 | - | 3 - 4 | 192 | 66 | |
FQ216715.1 | - | 3 - 4 | 210 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
