Assay Details
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007745.1 | NP_001007746.1 | 2 - 3 | 243 | 82 | |
GenBank mRNA | BC083626.1 | - | 2 - 3 | 243 | 82 | |
FQ221746.1 | - | 3 - 4 | 404 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
