Assay Details
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007745.1 | NP_001007746.1 | 4 - 5 | 479 | 63 | |
GenBank mRNA | BC083626.1 | - | 4 - 5 | 479 | 63 | |
FQ221746.1 | - | 5 - 6 | 640 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
